Dekglaasjes, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2975-246 2975-245 2975-223 2975-224 2975-225 2975-243 2975-244
CORN2975-246EA 401.1 EUR
CORN2975-246 CORN2975-245 CORN2975-223 CORN2975-224 CORN2975-225 CORN2975-243 CORN2975-244
Dekglaasjes, Corning®
Dekglaasjes
Special, optically clear, water-white, borosilicate cover glass is resistant to surface attachment or weathering and will remain clear for extended periods of time.
  • Ontworpen voor gebruik in algemene laboratoriummicroscopie
Niet-steriel.