Micro pipettes, disposable, PCR Pipettes
Leverancier: DRUMMOND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5-000-1001-X 5-000-1001-X10
DRUM5-000-1001-XEA 224 EUR
DRUM5-000-1001-X DRUM5-000-1001-X10
Micro pipettes, disposable, PCR Pipettes
Pipetten Micro-capillairen
Glass, disposable.
  • Positive displacement micro pipettes with stainless steel plungers
  • Measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
  • Decontaminate at temperature of 400 °F or higher