Catheter syringes
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6188 6187
SEMA6188EA 393 EUR
SEMA6188 SEMA6187
Catheter syringes
Spuiten Injectiespuiten, medisch
These catheter syringes have a PP barrel with silicone rubber piston.
  • Can be uesd as suction and irrigation syringes
  • Also used in technical purposes for pastes, glues and more
These syringes are gas sterilisable.
Verpakking: Individually packed.