Threaded adapters, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H978-40 H978-30
BOHLH978-40EA 23.9 EUR
BOHLH978-40 BOHLH978-30
Threaded adapters, BOLA
Fittingen en connectoren
Adaptors, made of PTFE, white, allow the use of BOLA multiple distributors for bottles with female thread GL 45 also on bottles with GL 32, GL 40 and S 40 threads.
  • Chemical resistant
  • Temperature resistant from −200 to +200 °C