Wash bottles, wide neck
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1164 1165
215-0255EA 25.1 EUR
215-0255 215-0256
Wash bottles, wide neck
Flessen Spuitflessen
LDPE, translucent
  • Wide neck for quick, safe filling, leakproof closure
  • Formed from a single piece, outlet tube can no longer become contaminated or leak. Jet tip can be cut to increase flow rate
  • Good chemical resistance, heat resistant up to +80 °C