Saccharometers
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2900FC000/20QP 2900FC005/20QP 2900FC010/20QP 2900FC020/20QP 2900FC030/20QP
614-0256EA 36.3 EUR
614-0256 614-0257 614-0258 614-0259 614-0260
Saccharometers
Hydrometers
These saccharometers are designed for reading below the meniscus.