Ultrapure water systems, Barnstead™ MicroPure™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
50132370 50132372 50132373 50132374
BARN50132370EA 7410 EUR
BARN50132370 BARN50132372 BARN50132373 BARN50132374
Ultrapure water systems, Barnstead™ MicroPure™
Waterzuiveringssystemen Puur- en ultrapuur-watersystemen
These water purification systems deliver ultrapure 18,2 MΩ.cm water with consistent quality. They are designed for laboratories that require up to 15 L/day of ultrapure water for HPLC or other instrumental analyses, sample preparation, glassware rinsing, or molecular biology work in which endotoxins and/or nucleases would interfere.
  • Optional integrated 6 L feed water reservoir: System can be placed without direct access to a water line
  • Validated 0,2 µM final filter can be autoclaved up to 5 times
  • Standard feed water monitor supports consistent water