Mouth mirrors
Leverancier: Aesculap
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DA026R DA027R DA036R DA037R
AESCDA026REA 33.3 EUR
AESCDA026R AESCDA027R AESCDA036R AESCDA037R
Mouth mirrors
Spiegels
The mouth mirror casts fine image without reflections under mid luminous conditions in the case of standard mirrors and high luminous conditions in the case of premium mirrors. The standard mirrors are ideal for use in standard treatments like scaling and the premium mirrors are ideal for surgery, treatment of deep cavities, endodontics and implantology.