Kidney dishes, Aesculap®
Leverancier: Aesculap
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JG504R JG506R JG508R
AESCJG504REA 0 EUR
AESCJG504R AESCJG506R AESCJG508R
Kidney dishes, Aesculap®
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
These dishesmade of stainless steel, can be used as instrument dishes and for a wide variety of other purposes.
  • Very good chemical resistance
  • Very good thermal resistance, autoclavable
  • Smooth surfaces for easy cleaning