Tubing connectors, T-shaped
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4657 4656
SEMA4657EA 8.3 EUR
SEMA4657 SEMA4656
Tubing connectors, T-shaped
Fittingen en connectoren
PP, natural
  • Temperature resistance up to +120 °C
  • Long conical connections for high tolerance ranges
  • Autoclavable