Closures for bottles, with seal ring, PP
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
466191 471003
SEMA466191EA 598 EUR
SEMA466191 SEMA471003
Closures for bottles, with seal ring, PP
Sluitingen Flesdoppen
PP closure, with PE sealing ring
  • Safety closures for Inter pharmaceutical glasses
  • Low water vapor permeability