Pipette fillers, Pip-Lab
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
901.001 901.003
ALLA901.001EA 17.9 EUR
ALLA901.001 ALLA901.003
Pipette fillers, Pip-Lab
Pipetten Pipetvullers
These manual pipette filler accept all types of pipettes thanks to their flexible connecting joint. They are ideal for use when pipetting corrosive or toxic liquids.
  • Easy to use even with one hand
  • Permits precise filling by turning the toothed wheel up or down
  • Enables easy decanting by pressing the button on the side.
  • Dismantles in seconds: all composant parts can be cleaned easily