Analytical HPLC columns, XSelect™ CSH C18
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
186005270 186005262
WATE186005270EA 0 EUR
WATE186005270 WATE186005262
Analytical HPLC columns, XSelect™ CSH C18
Kolommen voor chromatografie
XSelect™ CSH C18 is a general purpose column that provides alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology helps to overcome column related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed and slow mobile-phase re-equilibration.