VWR®, Ammonium (in N) 1000 mg/L in water standard for Ion Chromatography
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
84953.180EA
81.2
EUR
84953.180
84953.260
VWR®, Ammonium (in N) 1000 mg/L in water standard for Ion Chromatography
Standaarden
Kationische standaarden
Ammonium standaarden
Certificaten: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.