VWR®, Phosphate (in P) 1,000 mg/L in Water standard for ion chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
84989.180EA 72.8 EUR
84989.180 84989.260
VWR®, Phosphate (in P) 1,000 mg/L in Water standard for ion chromatography
Standaarden Anionische standaarden Fosfaat standaarden
Certificaten: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.