VWR® ARISTAR®, Potassium 1000 mg/L in Water Standard for Ion Chromatography
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
84990.180EA
64.4
EUR
84990.180
84990.260
VWR® ARISTAR®, Potassium 1000 mg/L in Water Standard for Ion Chromatography
Standaarden
Kationische standaarden
Kalium standaarden
Certificaten: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.