VWR® ARISTAR®, Sodium 1000 mg/L in Water Standard for Ion Chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
84993.180EA 64.4 EUR
84993.180 84993.260
VWR® ARISTAR®, Sodium 1000 mg/L in Water Standard for Ion Chromatography
Standaarden Kationische standaarden Natrium standaarden
Certificaten: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.