VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
84996.180EA 71.8 EUR
84996.180 84996.260
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Standaarden Anionische standaarden Sulfaat standaarden
Certificaten: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.