VWR®, Sulphite (in HSO³⁻) 1000 mg/L in Water standard for ion chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
84997.180EA 51.5 EUR
84997.180 84997.260
VWR®, Sulphite (in HSO³⁻) 1000 mg/L in Water standard for ion chromatography
Standaarden Anionische standaarden Sulfiet standaarden
Certificaten: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.