Dissecting scissors, Aesculap® Durotip®
Leverancier: Aesculap
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BC260R BC261R BC261W BC262R BC263R BC263W
AESCBC260REA 304 EUR
AESCBC260R AESCBC261R AESCBC261W AESCBC262R AESCBC263R AESCBC263W
Dissecting scissors, Aesculap® Durotip®
Scharen
Dissecting scissor for dental applications. Type Metzenbaum.
  • With inserted carbide cutting edges
  • Excellent cutting accuracy
  • Long working life
  • Gold-plated rings
Wave cut models prevent tissue from slipping out during the cutting process.