Sondes
Leverancier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1341-6331 1341-6332 1341-6335 1341-6337 1341-6338 1341-6339 1341-6340 1341-6341 1341-6336 1341-6332-0100
620-1860EA 62 EUR
620-1860 620-1861 620-2761 620-2394 620-2762 620-2763 620-2764 620-2765 620-2262 620-2823
Sondes
Temperatuursondes
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature.
Order Now
Specificaties
Environmentally Preferable