Laboratory coats, ZetDress®
Leverancier: Zellstoff-Vertriebs-GmbH & Co. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30813-01 30814-01
ZVGA30813-01EA 258 EUR
ZVGA30813-01 ZVGA30814-01
Laboratory coats, ZetDress®
Wegwerpkleding
These disposable coats are made of spunbond PP fleece. Ideal for use in sensitive work areas, such as the food processing industry and other hygienic environments.
  • With push buttons
  • Individually packaged
Colour: White