Lids, flat for Clifton stirred baths
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LD4-14 LD4-22 LD4-8
NICKLD4-14EA 264 EUR
NICKLD4-14 NICKLD4-22 NICKLD4-8
Lids, flat for Clifton stirred baths
Deksels Bath Lids
Stainless steel flat lids.
  • The use of a lid helps reduce evaporation and assists in keeping samples free from contamination
  • Insulated handles