Temperature indicating strips
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TC8020 TC8010 TC5050 TC5045 TC5040 TC5020 TC5025 TC5030 TC5035 TC8030 TC8040 TC8050
SIMETC8020EA 32.1 EUR
SIMETC8020 SIMETC8010 SIMETC5050 SIMETC5045 SIMETC5040 SIMETC5020 SIMETC5025 SIMETC5030 SIMETC5035 SIMETC8030 SIMETC8040 SIMETC8050
Temperature indicating strips
Temperatuurindicatiestrips
These temperature indicating strips are designed for use in electronics, food and pharmaceutical industries, and also in the oven and engine construction businesses. They are particularly useful for indicating temperatures of moving parts.
  • Self adhesive foils to measure maximum surface temperatures
  • Indicators change colour when marked temperature is reached and the change is irreversible
  • Highest temperature remains visible for months
Leveringsinformatie: Series 5 and Series 8 contains 10 stickers per sheet.