Universal replacement parts, Omnifit®
Leverancier: OMNIFIT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006FR-06-05 006FR-06-10 006FR-06-25-PE 006FR-10-05 006FR-10-10 006FR-10-20 006FR-10-25-PE 006FR-15-05 006FR-15-10 006FR-15-25-PE 006FR-25-05 006FR-25-10 006FR-25-25-PE 006FR-35-05 006FR-35-10 006FR-35-25-PE 006FR-50-05 006FR-50-10 006FR-50-25-PE 006OR-10-V 006FR-10-50
OMNI006FR-06-05EA 64.4 EUR
OMNI006FR-06-05 OMNI006FR-06-10 OMNI006FR-06-25-PE OMNI006FR-10-05 OMNI006FR-10-10 OMNI006FR-10-20 OMNI006FR-10-25-PE OMNI006FR-15-05 OMNI006FR-15-10 OMNI006FR-15-25-PE OMNI006FR-25-05 OMNI006FR-25-10 OMNI006FR-25-25-PE OMNI006FR-35-05 OMNI006FR-35-10 OMNI006FR-35-25-PE OMNI006FR-50-05 OMNI006FR-50-10 OMNI006FR-50-25-PE OMNI006OR-10-V OMNI006FR-10-50
Universal replacement parts, Omnifit®
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Universal replacement parts for all Omnifit® labware columns which gives complete flexibility to replace lost or worn parts. Compatible with BenchMark, SolventPlus and HiT column ranges.