Flame photometers, 360
Leverancier: SHERWOOD SCIENTIFIC INSTRUMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36000009 36000209
SHEW36000009EA 4380 EUR
SHEW36000009 SHEW36000209
Flame photometers, 360
Spectrofotometers en fotometers Vlamfotometers
Single channel flame photometer for analysis of five elements (sodium, potassium, lithium, calcium, barium) designed with front loading, unobstructed sample work area and removable tray for easy cleaning.
  • Automatic ignition and flame optimisation
  • Safety features including fail safe flame detection and shut off with low air pressure indicator
  • High sensitivity with fine manual control of air and stable flame design
  • Indicator on the front panel indicate the filter selected and the flame status
  • Useful display 'HOLD' button to ensure that readings are not lost