Spectroscopy cuvettes
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B0631070 B0631071
PERCB0631070EA 186 EUR
PERCB0631070 PERCB0631071
Spectroscopy cuvettes
Cuvetten Cuvetten voor spectrofotometers
These high quality cuvettes allows you to make full use of the accuracy and sensitivity of your optical instrument.
  • High transmission
  • Accurate pathlength
  • Windows are fused to the bodies to avoid any deformation