Thermometer Probe, TPN
Leverancier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1341-0646 1343-1005 1341-1000 1341-0800 1343-0800
EBRO1341-0646EA 83 EUR
EBRO1341-0646 620-0232 EBRO1341-1000 EBRO1341-0800 EBRO1343-0800
Thermometer Probe, TPN
Temperatuursondes
The flexible thermal wire probes are used to measure air, oven and glass temperatures.