Mopping system Clino®, with special mops, for cleanroom cleaning
Leverancier: PFENNIG REINIGUNGSTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3500902 2700266 2700268
141-1039EA 186 EUR
141-1039 141-1034 141-1035
Mopping system Clino®, with special mops, for cleanroom cleaning
Moppen
Mops made from 100% polyester microfibre yarn (0,5 De)
  • Suitable for cleaning and disinfection of GMP areas
  • Maximum efficiency due to incorporated microfibres
  • Washable and autoclavable (up to 50 treatment cycles)
  • For flat wringer Ringo CR