Handheld digital microscopes, Dino-Lite Medical, DermaScope®
Leverancier: Dino-Lite / IDCP B.V.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MEDL7DM MEDL4DW MEDL7DW
630-2617EA 908 EUR
630-2617 630-2511 IDCPMEDL7DW
Handheld digital microscopes, Dino-Lite Medical, DermaScope®
Microscopen
These handy, easy-to-use microscopes can be quickly deployed and used to create sharp and clear photos and videos of a wide variety of skin problems.
  • Without pain or long waiting times for the patient
  • Built-in adjustable polariser reduces the gloss effects of the skin
  • Support the use of water or oil (not always necessary)
  • Illumination: 8 white LEDs (switchable)
  • High speed USB 2.0 output
  • HR models come with a metal housing
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
Ideal for physicians, dermatologists or skin specialists.
Certificaten: Medical Device Directive 93/42/CEE modified 2007/47/EC.
Leveringsinformatie: Supplied with a protecting pouch, software and calibration target.