Adhesive qPCR film seals, QuickSeal qPCR Crystal™
Leverancier: iST Scientific Ltd
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IST-121-080LR IST-121-080LS IST-121-080SR IST-121-080SS IST-121-080TS
731-0451EA 777 EUR
731-0451 731-0452 731-0453 731-0454 731-0455
Adhesive qPCR film seals, QuickSeal qPCR Crystal™
Afdichtingen Afdichtingen van microtiterplaten
Clear, non-tacky, pressure sensitive film for qPCR plates (96- or 384-well) and situations where fluorescence is experienced.
  • Optically clear, minimal auto-fluorescence
  • DMSO resistant
  • Non-pierceable, peelable
  • Seal integrity range: −40 to +110 °C
  • Available sterile or non sterile