Bodywarmer, Technik™, TK51
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TK51BKRXL TK51BKRXXL TK51BKRM TK51BKRS TK51BKRL
PRTWTK51BKRXLEA 64 EUR
PRTWTK51BKRXL PRTWTK51BKRXXL PRTWTK51BKRM PRTWTK51BKRS PRTWTK51BKRL
Bodywarmer, Technik™, TK51
Werkkleding
This bodywarmer is constructed from 94% polyester and 6% Spandex, laminated with 100% polyester micro fleece for extra warmth and comfort. The softshell material is waterproof, windproof and breathable.
  • 3-layer laminated bodywarmer with collar
  • Lightweight alternative to a jacket
  • Ideal for more active working environments
  • Waterproof zips
  • One breast pocket with zip closure
Weight: 310 g/m²