Self-wringing cleanroom roller mops, Roll-O-Matic®
Leverancier: FHP VILEDA SCS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
118771 120848 121165 119654 119657 119658 118550 118549 122910
129-0195EA 203 EUR
129-0195 129-0270 129-0271 VILE119654 VILE119657 VILE119658 141-1440 141-1439 141-1441
Self-wringing cleanroom roller mops, Roll-O-Matic®
Moppen
Roll-O-Matic® is the ideal tool for cleaning or disinfecting floors, walls and ceilings in hygiene-sensitive areas.
It has a self-wringing mechanism, and is best used with the 'pull and lift' technique recommended for cleaning and disinfecting. It’s simple to use, and elminates bulky, expensive wringers.
  • Integrated wringing mechanism for use with 'pull and lift' technique
  • Coloured clips on the handles allow for colour coding of equipment
  • Stainless steel handles are compatible with gamma and autoclave sterilisation
  • Straight head design allows the user to flip-over the mop head to use both sides
Aluminium and stainless steel Roll-O-Matic® mops are ideal when seeking one mop to apply disinfectant, clean surfaces, or absorb liquids in production and controlled environments. Aluminium extendable and stainless steel Roll-O-Matic® mops are constructed to resist repeated autoclave cycles, and continually being exposed to common chemicals and disinfectants used in controlled environments.
Roll-O-Matic® mop head refills feature new foam, manufactured specifically for the controlled environments, that allows 25% more liquid absorption and 2× more liquid release than similar foams. Completely autoclavable, resistant to gamma and ETO sterilisation. Each refill is individually packed and lot number controlled.