Cleaning trolleys, Clino® CR3 / CR5, with flat wringer
Leverancier: PFENNIG REINIGUNGSTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3570000 3530010
141-1044EA 3410 EUR
141-1044 141-1041
Cleaning trolleys, Clino® CR3 / CR5, with flat wringer
Transportwagens
For areas with the highest requirements on GMP conformity. Made entirely of stainless steel.
  • Convenient for the use in cleanrooms
  • GMP-conformity
  • Complete unit can be autoclaved
  • Highly efficient cleaning and disinfection
  • Assured cross-contamination prevention
  • Object-specific adjustments are possible
  • Flat mopping allows a significant reduction of working time with a much better ergonomic work process