Container, EUROBOX
Leverancier: ZARGES LEICHTBAU
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40700 40706 40702 40709
ZARG40700EA 0 EUR
ZARG40700 ZARG40706 ZARG40702 ZARG40709
Container, EUROBOX
Dozen Opbergdozen
Aluminium, with lid.
  • Lid with stainless steel hinges and holding straps
  • Blue stacking corners ensure safe stacking
  • Corner beads for additional dimensional stability
Eurobox has high resistance against corrosion due to stainless steel fittings. The peripheral foamed seal in the lid protects against dust and splash water.