HPLC columns, LC-ABZ, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
59142 59142C40 59140-U 59544-U 59545-U 59610 59611 59141 59142C30
59142.EA 958 EUR
59142. 59142C40 59140-U 59544-U 59545-U 59610. 59611. 59141. 59142C30
HPLC columns, LC-ABZ, Supelco®
Kolommen voor chromatografie
The LC-ABZ phase has a unique deactivation technology which provides excellent reversed-phase performance for basic compounds, as well as those that are acidic, polar neutral, and non-polar.