Alcohol meters
Leverancier: GERING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0771WG 0772WG 0773WG 0775WG 0776WG 0777WG 0778WG 0779WG 0780WG 0781WG 0782WG 0783WG 0784WG 0785WG 0786WG 0787WG 0788WG 0789WG 0768WG 0750WG
GERI0771WGEA 0 EUR
GERI0771WG GERI0772WG GERI0773WG GERI0775WG GERI0776WG GERI0777WG GERI0778WG GERI0779WG GERI0780WG GERI0781WG GERI0782WG GERI0783WG GERI0784WG GERI0785WG GERI0786WG GERI0787WG GERI0788WG GERI0789WG 614-0567 GERI0750WG
Alcohol meters
Hydrometers
Alcohol meters with a standard temperature of 20 °C.
Certificaten: DIN 12803
Bestelinformatie: Includes a thermometer.