SPE plates, 96-well, HyperSep™, C18
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60300-428 60300-425 60300-426 60300-427 60307-204
549-0671EA 485 EUR
549-0671 549-0668 549-0669 549-0670 HYPE60307-204
SPE plates, 96-well, HyperSep™, C18
Microplaten Platen voor vastefase-extractie
Thermo Scientific HyperSep™ C18 SPE 96-well plates and individual wells feature a highly retentive alkyl-bonded phase for non polar to moderately polar compounds. The hydrophobic reversed phase material is retentive for most nonpolar compounds, and retains most organic analytes from aqueous matrices.
  • For many biological and pharmaceutical applications
  • Retains most organic analytes