Hot plates, SG, SGR and SGK series
Leverancier: GESTIGKEIT HARRY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SGK1 SGK6 SG3 SG5 SGR3 SGK4 SGR4 SG2 SG6 SGR2
GESTSGK1EA 408 EUR
GESTSGK1 GESTSGK6 GESTSG3 GESTSG5 GESTSGR3 GESTSGK4 GESTSGR4 GESTSG2 GESTSG6 GESTSGR2
Hot plates, SG, SGR and SGK series
Verwarmers Verwarmingsplaten
These hotplates made from smoothly finished cast-iron have a concave central expansion recesses. They are used for heating units which have a screw-in fitting for a supporting retort rod and have been designed for continuous operation .