Precision laboratory thermometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1042371 1042372 1042381 1042373 1042374 1042375 1042380 1042376 1042377 1042382 1042378 1042379 1042383 1042341 1042342 1042350 1042343 1042344 1042345 1042351 1042346 1042347 1042348 1042349 1042354 1042353 1042384 1042343S 1042344S 1042345S 1042351S 1042346S 1042604S 1042347S 1042349S/04/3P 1042352S 1042703S/04/3P 1042703S 1042704S 1042353S 1042349S 1042354S 1122904S 1122905S 1122906S 1122907S 1042343S/03/3P 1042344S/03/3P 1042345S/03/3P 1042351S/03/3P 1042604S/03/3P 1042346S/03/3P 1042347S/03/3P 1042352S/03/3P 1042703S/03/3P 1042704S/03/3P 1042353S/03/3P 1042349S/03/3P 1042354S/03/3P 1122904S/03/3P 1122905S/03/3P 1122906S/03/3P 1122907S/03/3P 1042343S/04/3P 1042344S/04/3P 1042345S/04/3P 1042351S/04/3P 1042604S/04/3P 1042346S/04/3P 1042347S/04/3P 1042352S/04/3P 1042704S/04/3P 1042353S/04/3P 1042354S/04/3P 1122904S/04/3P 1122905S/04/3P 1122906S/04/3P 1122907S/04/3P 1042381/04/3P
SCNL1042371EA 33 EUR
SCNL1042371 SCNL1042372 SCNL1042381 SCNL1042373 SCNL1042374 SCNL1042375 SCNL1042380 SCNL1042376 SCNL1042377 SCNL1042382 SCNL1042378 SCNL1042379 SCNL1042383 SCNL1042341 SCNL1042342 SCNL1042350 SCNL1042343 SCNL1042344 SCNL1042345 SCNL1042351 SCNL1042346 SCNL1042347 SCNL1042348 SCNL1042349 SCNL1042354 SCNL1042353 SCNL1042384 SCNL1042343S SCNL1042344S SCNL1042345S SCNL1042351S SCNL1042346S SCNL1042604S SCNL1042347S SCNL1042349S/04/3P SCNL1042352S SCNL1042703S/04/3P SCNL1042703S SCNL1042704S SCNL1042353S SCNL1042349S SCNL1042354S SCNL1122904S SCNL1122905S SCNL1122906S SCNL1122907S SCNL1042343S/03/3P SCNL1042344S/03/3P SCNL1042345S/03/3P SCNL1042351S/03/3P SCNL1042604S/03/3P SCNL1042346S/03/3P SCNL1042347S/03/3P SCNL1042352S/03/3P SCNL1042703S/03/3P SCNL1042704S/03/3P SCNL1042353S/03/3P SCNL1042349S/03/3P SCNL1042354S/03/3P SCNL1122904S/03/3P SCNL1122905S/03/3P SCNL1122906S/03/3P SCNL1122907S/03/3P SCNL1042343S/04/3P SCNL1042344S/04/3P SCNL1042345S/04/3P SCNL1042351S/04/3P SCNL1042604S/04/3P SCNL1042346S/04/3P SCNL1042347S/04/3P SCNL1042352S/04/3P SCNL1042704S/04/3P SCNL1042353S/04/3P SCNL1042354S/04/3P SCNL1122904S/04/3P SCNL1122905S/04/3P SCNL1122906S/04/3P SCNL1122907S/04/3P SCNL1042381/04/3P
Precision laboratory thermometers
Thermometers Analoge thermometers
These total immersion thermometers are suitable for official certification.
  • Colourless prismatic capillary form
Red or blue filling, enclosed or stem type.
Bestelinformatie: Additional thermometer models are also available (separately) with works certificate or with DAkkS calibration certificate (see bottom of ordering table).