In-line filter cartidges, low pressure
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-CFE-S75-5 JR-CFE-S2-5 JR-CFE-S10-5
JOURJR-CFE-S75-5EA 0 EUR
JOURJR-CFE-S75-5 JOURJR-CFE-S2-5 JOURJR-CFE-S10-5
In-line filter cartidges, low pressure
Filters Filterpatronen
Filters are constructed of PTFE and CTFE, with a type 316 stainless low pressure drop screen. The inner design of the cartridge ensures the equal distribution of the solvent to the screen.
  • Compact design and easy to replace
  • Fit all 1/4 - 28 fitting details
  • Three porosity options