Precision density hydrometers, M100
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3192005 3192006 3192014 3192013 3192012 3192011 3192010 3192009 3192008 3192004 3192003 3192002 3192001 3192007
SCNL3192005EA 58.9 EUR
SCNL3192005 SCNL3192006 SCNL3192014 SCNL3192013 SCNL3192012 SCNL3192011 SCNL3192010 SCNL3192009 SCNL3192008 SCNL3192004 SCNL3192003 SCNL3192002 SCNL3192001 SCNL3192007
Precision density hydrometers, M100
Hydrometers
These precision density hydrometers without thermometers are suitable for calibration.
Error limit: ±0,002 g/cm³
Reference temperature: 20 °C
Certificaten: DIN 12791
Bestelinformatie: Surface tension in class M (medium) on request.
Verpakking: Individually in a square plastic sleeve.