Mask carriers, A 189 series
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A18940-09-0715 A18925-09-0715 A18970-15-0715 A18950-12-0715 A058-50-0515 A058-70-0515 A18920090515 A18960250715_U
ENTEA18940-09-0715EA 0 EUR
ENTEA18940-09-0715 ENTEA18925-09-0715 ENTEA18970-15-0715 ENTEA18950-12-0715 ENTEA058-50-0515 ENTEA058-70-0515 ENTEA18920090515 ENTEA18960250715_U
Mask carriers, A 189 series
Wafertransportsystemen Wafer dragers
These carriers are designed to hold masks or reticles during transport. They can be used within diverse industries, such as semi-conductor data storage, chemical processing and fuel cell. The carriers can be combined with handles and/or shipping boxes (need to be ordered separately).