Wafer carrier handles for 125 mm (5") wafer handling
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A72-50-04-0215 A72-50-03-0215 A040-101-0215 A047-0215 A725004B0215
216-1633EA 0 EUR
216-1633 ENTEA72-50-03-0215 ENTEA040-101-0215 ENTEA047-0215 ENTEA725004B0215
Wafer carrier handles for 125 mm (5") wafer handling
Wafertransportsystemen Wafer dragers
Squeeze style.
  • Attaches to the notches on both end walls of carrier
  • Overall size: 163×140×183 mm
  • Compatible with all 'M' style 182 series, 194 series, 72 series, 190 series wafer carriers
Squeeze style
Attaches to the notches on both end walls of carrier
Overall size: 165×130×165 mm
Compatible with all 'M' style 182 series, 194 series, 72 series wafer carriers
Squeeze style with support bar
Attaches to the notches on both end walls of carrier
Overall size: 165×152×203 mm
Compatible with all 'M' style 182 series, 194 series, 72 series wafer carriers
Slingshot style
Slides onto the top flanges of carrier
Overall size: 15×170×246 mm
Articles may not be available in all countries. Please contact your local Avantor office.