Bottles, wide mouth, square, with pressure seal cap
Leverancier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
55 539 58
LAMA55EA 30.1 EUR
LAMA55 LAMA539 LAMA58
Bottles, wide mouth, square, with pressure seal cap
Flessen Kunststof flessen
HDPE, translucent.
  • For food use
  • With self sealing tamper evident pressure cap
The square shape allows a better use of the spaces. The self-sealing screwing cap is tamper proof. Thanks to the wide mouth they are suitable for the storage of creams, semi-solid substances, granulates and powders.