Magnetic stirrer heads with conical joint
Leverancier: JUCHHEIM Laborgerate GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10262.00 10264.00
JUCH10262.00EA 1300 EUR
JUCH10262.00 JUCH10264.00
Magnetic stirrer heads with conical joint
Roerkoppen
Conical joint stirrer heads made of stainless steel bottom, special steel shaft, chromium plated brass upper body for vacuum and slight positive pressure.