Capillary columns, TRACE™ TR-Biodiesel
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26AA395P 26AF024P 26AX142P
555-9078EA 853 EUR
555-9078 555-9076 555-9129
Capillary columns, TRACE™ TR-Biodiesel
Kolommen voor chromatografie
These columns are designed for use in carbon neutral fuels development applications.
TR-BioDiesel (M) columns: For determination of residual methanol content in Biodiesel according to EN method 14110.
TR-BioDiesel (G) columns: For analysis of total glyceride content according to EN method 14105.
TR-BioDiesel (F) columns: For analysis of FAME content in biodiesel according to EN method 14103.
TR-BioDiesel (G) ASTM columns: For analysis of total glyceride content according to ASTM method D-6584.