Inlet filter, Rheodyne®
Leverancier: IDEX RHEODYNE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7315-010 7335-010
554-2127EA 341 EUR
554-2127 554-2129
Inlet filter, Rheodyne®
Filters Filters voor vloeistofverwerking
Inlet filters are used to extract particles from the sample between the injector and the column.