HPLC columns, MonoChrom® Si
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A0501250X212 A0511050X046 A0511150X046
VARIA0501250X212EA 0 EUR
VARIA0501250X212 VARIA0511050X046 VARIA0511150X046
HPLC columns, MonoChrom® Si
Kolommen voor chromatografie
MonoChrom® columns provide excellent chromatography of organic amines, organic acids, amphoteric compounds and neutral compounds.