Flame photometric detectors
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19256-60700 19256-80010 19256-80030 5061-5886 5061-5890 0905-1610 19256-60800 2190-0584
HEWL19256-60700EA 183 EUR
HEWL19256-60700 HEWL19256-80010 HEWL19256-80030 HEWL5061-5886 HEWL5061-5890 HEWL0905-1610 HEWL19256-60800 HEWL2190-0584
Flame photometric detectors
Chromatografie detectoren
FPD is designed with minimum detectable levels of 3,6 pg/s for sulfur and 60 fg/s for phosphorus.
Bestelinformatie: Upgrade kits are available and can be ordered separately.